missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin hydrochloride, MedChemExpress

Product Code. 16498398
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498398 10 mg 10mg
16484287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498398 Supplier Medchem Express Supplier No. HYB0192A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).

Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).

Specifications

Chemical Name or Material Alfuzosin hydrochloride
CAS 81403-68-1
Color White
Purity Grade Notes Research
Molecular Formula C19H28ClN5O4
Quantity 10 mg
Synonym SL 77499-10
Solubility Information H2O : 50 mg/mL (117.40 mM; Need ultrasonic) ∣DMSO : 31.25 mg/mL (73.37 mM; Need ultrasonic)
SMILES O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C.Cl
Molecular Weight (g/mol) 425.91
Formula Weight 425.91
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.