missing translation for 'onlineSavingsMsg'
Learn More

(±)-Alfuzosin-d7 HCl (tetrahydrofuroyl-d7), TRC

Product Code. 30323623
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323623 5 mg 5mg
30311773 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30323623 Supplier Toronto Research Chemicals Supplier No. TRCA5320015MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alfuzosin-D7 (tetrahydrofuranyl-D7) Hydrochloride
CAS 1276197-14-8
Molecular Formula C19 D7 H20 N5 O4 . Cl H
Quantity 5 mg
Synonym N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]-2,3,3,4,4,5,5-heptadeuteriooxolane-2-carboxamide hydrochloride, (±)-Alfuzosin-d7 HCl (tetrahydrofuroyl-d7), N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]tetrahydro-2-furancarboxamide-2,3,3,4,4,5,5-d7 hydrochloride, Alfuzosin-d7 hydrochloride, Alfuzosin-D7 (tetrahydrofuranyl-D7) hydrochloride, (2RS)-N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]tetrahydrofuran-2-carboxamide-2,3,3,4,4,5,5-d7 hydrochloride
InChI Formula InChI=1S/C19H27N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h10-11,14H,4-9H2,1-3H3,(H,21,25)(H2,20,22,23);1H/i4D2,6D2,9D2,14D;
SMILES Cl.[2H]C1([2H])OC([2H])(C(=O)NCCCN(C)c2nc(N)c3cc(OC)c(OC)cc3n2)C([2H])([2H])C1([2H])[2H]
IUPAC Name N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]-2,3,3,4,4,5,5-heptadeuteriooxolane-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 432.9529
Formula Weight 432.2269
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.