missing translation for 'onlineSavingsMsg'
Learn More

Alflutinib mesylate, MedChemExpress

Product Code. 16491007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491007 50 mg 50mg
16477437 5 mg 5mg
16455546 100 mg 100mg
16434728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491007 Supplier Medchem Express Supplier No. HY112870A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Alflutinib (Furmonertinib) mesylate is is a potent inhibitor of EGFR. Alflutinib (Furmonertinib) mesylate inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Alflutinib (Furmonertinib) mesylate has the potential for the research of cancer diseases, especially non-small cell lung cancer (NSCLC).

Alflutinib (Furmonertinib) mesylate is is a potent inhibitor of EGFR. Alflutinib (Furmonertinib) mesylate inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Alflutinib (Furmonertinib) mesylate has the potential for the research of cancer diseases, especially non-small cell lung cancer (NSCLC).

Specifications

Chemical Name or Material Alflutinib mesylate
CAS 2130958-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H35F3N8O5S
Quantity 50 mg
Synonym Furmonertinib mesylateAST2818 mesylate
Solubility Information DMSO : 5 mg/mL (7.52 mM; Need ultrasonic)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OCC(F)(F)F)N=C1N(CCN(C)C)C)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 664.7
Formula Weight 664.7
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.