missing translation for 'onlineSavingsMsg'
Learn More

Alflutinib mesylate, MedChemExpress

Product Code. 16455546
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455546

Brand: Medchem Express HY112870A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alflutinib (Furmonertinib) mesylate is is a potent inhibitor of EGFR. Alflutinib (Furmonertinib) mesylate inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Alflutinib (Furmonertinib) mesylate has the potential for the research of cancer diseases, especially non-small cell lung cancer (NSCLC).

Alflutinib (Furmonertinib) mesylate is is a potent inhibitor of EGFR. Alflutinib (Furmonertinib) mesylate inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Alflutinib (Furmonertinib) mesylate has the potential for the research of cancer diseases, especially non-small cell lung cancer (NSCLC).

Specifications

Chemical Name or Material Alflutinib mesylate
CAS 2130958-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H35F3N8O5S
Quantity 100 mg
Synonym Furmonertinib mesylateAST2818 mesylate
Solubility Information DMSO : 5 mg/mL (7.52 mM; Need ultrasonic)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OCC(F)(F)F)N=C1N(CCN(C)C)C)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 664.7
Formula Weight 664.7
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.