Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
Product Code. 11365497
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365497 5 kg 5kg
11375497 50 g 50g
11385497 250 g 250g
11395497 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11365497 Supplier Thermo Scientific Chemicals Supplier No. 042726.A7
Request Bulk or Custom Formats
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size -100/200 Mesh
Pore Size 150 Å
Packaging Drum
pH 6.5 to 7.5
Quantity 5 kg
Surface Area (Metric) 350 to 400 m2/g
Type Silica Gel
Material Silica