missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ USA Standard Sieve, 80 Mesh
Brass frame and Stainless-steel mesh sieves
Brand: Thermo Scientific™ 039986.NK
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
- Sieves are full height
- Mesh pores: 177μ ( 0.177mm )
- 3 in. diameter sieves are 1 in. deep
- 8 in. diameter sieves are 2 in. deep
- 12 in.diameter sieves are 3 in. deep
Specifications
80 Mesh | |
3 in. | |
Sieve | |
12 in. |
30.4cm | |
7.62cm | |
ASTM E11 | |
Stainless Steel |
RUO – Research Use Only