missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Wide range of working temperatures with upper limit of 290°C
- Remains rigid if heated above notional melting point of 327°C
- Do not melt or deform if overheated until 400°C
- Non-stick with opaque clarity
Specifications
0.81mm (0.031 in.) thick | |
PTFE | |
PTFE sheet |