Learn More
Thermo Scientific Chemicals ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip, 7.5-14 pH range
Product Code. 11313607
Change view
Click to view available options
pH Range:
5 to 10
Quantity:
1 Unit
6 Unit
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Quantity unitSize
11313607 5 to 10 6 Unit 6 units
11323607 5 to 10 1 Unit 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11313607 Supplier Thermo Scientific Chemicals Supplier No. 035234.LS
Request Bulk or Custom Formats
ColorpHast™ pH Strips, Universal, four squares with different indicators on each strip, 0-14 pH range is used in analytical laboratory to measure the pH readings in seconds in drinking water, saliva and in chemical reactions. It is utilized for quick and accurate measurement and saves our time and trouble.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
pH Range 5 to 10
Quantity 6 Unit
Type Intermediate pH Strip