Learn More
Amberlite™ IRA-200C(Na), ion exchange resin
Description
Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Amberlite IRA-200C(Na) |
| Physical Form | Beads |
| Odor | Odorless |
| Molecular Formula | Styrene-DVB |
| MDL Number | MFCD01864001 |
| Density | 1.1-1.4g/cm3 |
| Color | Amber |
| Quantity | 5 kg |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.