missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-200C(Na), ion exchange resin

Product Code. 11359907
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359907 1 kg 1kg
11379907 250 g 250g
11369907 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359907 Supplier Thermo Scientific Alfa Aesar Supplier No. 042252.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Styrene-DVB; Strongly acidic macroreticular cation exchange resin | Styrene-DVB

Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amberlite IRA-200C(Na)
Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD01864001
Color Amber
Quantity 1 kg
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.