missing translation for 'onlineSavingsMsg'
Learn More

Alexidine Dihydrochloride, TRC

Product Code. 30006651
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006651 50 mg 50mg
30012046 500 mg 500mg
30011775 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006651 Supplier Toronto Research Chemicals Supplier No. TRCA52500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1715-30-6
Formula Weight 580.4223
InChI Formula InChI=1S/C26H56N10.2ClH/c1-5-9-15-21(7-3)19-33-25(29)35-23(27)31-17-13-11-12-14-18-32-24(28)36-26(30)34-20-22(8-4)16-10-6-2;;/h21-22H,5-20H2,1-4H3,(H5,27,29,31,33,35)(H5,28,30,32,34,36);2*1H
IUPAC Name 1-(2-ethylhexyl)-3-[N-[6-[[N-[N-(2-ethylhexyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]guanidine;dihydrochloride
Molecular Formula C26 H56 N10 . 2 Cl H
Molecular Weight (g/mol) 581.71
SMILES Cl.Cl.CCCCC(CC)CNC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)NCC(CC)CCCC
Chemical Name or Material Alexidine, Dihydrochloride

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.