missing translation for 'onlineSavingsMsg'
Learn More

Alexa Fluor™ 647 Anti-Cytokeratin 14 antibody [LL002], Abcam

Product Code. 32097762 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32097762 10 μL 10µL
32097961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32097762 Supplier Abcam Supplier No. ab32178310uL
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Knockout Tested Mouse Monoclonal Cytokeratin 14 antibody - conjugated to Alexa Fluor™ 647. Suitable for ICC/IF, IHC-P and reacts with Human samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Keratin, type I cytoskeletal 14 (Cytokeratin 14)
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE)
Cellular Localization Cytoplasm; Nucleus; Expressed in both as a filamentous pattern.
Classification Monoclonal
Clone LL002
Conjugate Alexa Fluor 647
Formulation pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 1% BSA
Gene KRT14
Gene Accession No. P02533
Host Species Mouse
Immunogen Keratin, type I cytoskeletal 14 (Cytokeratin 14)
Purification Method Affinity purification
Quantity 10 μL
Regulatory Status RUO
Research Discipline Oncology
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG3
Shipping Condition Blue Ice
Target Gene KRT14
Target Molecule Keratin, type I cytoskeletal 14 (Cytokeratin 14)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.