missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 594 Tyramide SuperBoost™ Kit, goat anti-mouse IgG
Click to view available options
Quantity:
Sufficient for 150 Slides
Sufficient for 50 Slides
Unit Size:
Pack of 150
Pack of 50
Description
SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).
Features:
- High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
- Simple protocol and detection using standard filters
- Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
- Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
- Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
- Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
- Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
- Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
- Stable for 6 months after receiving
Specifications
Specifications
| Conjugate | Alexa Fluor 594 |
| Quantity | Sufficient for 50 Slides |
| Content And Storage | 1 kit sufficient for 50 microscope slides (18 x 18 mm), containing: Blocking buffer (1X), 22.5 mL |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Product Line | SuperBoost |
| Product Type | Tyramide Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction