missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Streptavidin, 10 nm colloidal gold conjugate

Product Code. 10674603
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10674603

Brand: Invitrogen™ A32361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Alexa Fluor™ 488 Streptavidin, 10 nm colloidal gold conjugate

Alexa Fluor™ 488 Streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 488 Streptavidin Conjugates:
  • Alexa Fluor™ 488 streptavidin conjugate has Ex/Em maxima of ∼ (495/519)
  • Bright, photostable fluorescence
  • High solubility in aqueous solutions
  • Available in multiple colors
  • Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

    Properties of Alexa Fluor™ Dyes
    Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

    Blocking Endogenous Biotin
    Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 30 μg/mL
    Conjugate Alexa Fluor 488, Colloidal Gold
    Quantity 0.5 mL
    Content And Storage Store in refrigerator (2–8°C) and protect from light.
    Shipping Condition Wet Ice
    Form Liquid
    Product Line Alexa Fluor™
    Product Type Streptavidin Conjugate (fluorescent)
    Label or Dye Alexa Fluor Dyes

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.