Learn More
Alendronate sodium trihydrate, 97%
Description
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.
Solubility
Soluble in water at 10mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Merck Index | 14,229 |
| Solubility Information | Soluble in water at 10mg/ml |
| Formula Weight | 325.12 (271.08 Anhydrous) |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | Alendronate sodium trihydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.