missing translation for 'onlineSavingsMsg'
Learn More

Alendronate sodium hydrate, MedChemExpress

Product Code. 16475286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475286 100 mg 100mg
16461596 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475286 Supplier Medchem Express Supplier No. HY11101
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alendronate (sodium hydrate) is a farnesyl diphosphate synthase inhibitor with IC50 of 460 nM.

Alendronate (sodium hydrate) is a farnesyl diphosphate synthase inhibitor with IC50 of 460 nM.

Specifications

Chemical Name or Material Alendronate sodium hydrate
Physical Form Solid
CAS 121268-17-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C4H18NNaO10P2
For Use With (Application) Cancer-programmed cell death
Synonym Alendronate MK 217 G-704650 Adronat
Solubility Information H2O : ≥ 28.57 mg/mL (87.88 mM)
SMILES OP(C(O)(CCCN)P(O)(O[Na])=O)(O)=O.[3H2O]
Molecular Weight (g/mol) 325.12
Formula Weight 325.12
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.