missing translation for 'onlineSavingsMsg'
Learn More

Alectinib, MedChemExpress

Product Code. 16481056
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481056 200 mg 200mg
16465478 10 mg 10mg
16456066 100 mg 100mg
16431657 50 mg 50mg
16418197 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481056 Supplier Medchem Express Supplier No. HY13011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alectinib (CH5424802) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.

Alectinib (CH5424802) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.

Specifications

Chemical Name or Material Alectinib
CAS 1256580-46-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H34N4O2
Quantity 200 mg
Synonym CH5424802 RO5424802 AF802
Solubility Information DMSO : 6.2 mg/mL (12.85 mM; Need warming)
SMILES N#CC1=CC2=C(C3=C(N2)C(C)(C4=CC(N5CCC(CC5)N6CCOCC6)=C(C=C4C3=O)CC)C)C=C1
Molecular Weight (g/mol) 482.62
Formula Weight 482.62
Percent Purity 98.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.