missing translation for 'onlineSavingsMsg'
Learn More

Aldosterone (Hemiacetal) (1.0mg/mL in Acetonitrile), TRC

Product Code. 30180040
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180040 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180040 Supplier Toronto Research Chemicals Supplier No. TRCKIT03001X1ML
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52-39-1
Molecular Formula C21 H28 O5
Quantity 1 x 1 mL
Synonym Pregn-4-en-18-al, 11,21-dihydroxy-3,20-dioxo-, (11β)-, Aldosterone (8CI), (+)-Aldosterone, 11β,21-Dihydroxy-3,20-diketo-4-pregnen-18-al, 11β,21-Dihydroxy-3,20-diketopregn-4-ene-18-al, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione, 18-Formyl-11β,21-dihydroxy-4-pregnene-3,20-dione, 18-Oxocorticosterone, Aldocorten, Aldocortene, Aldocortin, Electrocortin, Elektrocortin, NSC 73856, Reichstein X, d-Aldosterone
InChI Formula InChI=1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,11,14-17,19,22,25H,2-7,9-10H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O
IUPAC Name (8S,9S,10R,11S,13R,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-13-carbaldehyde
Molecular Weight (g/mol) 360.44
Formula Weight 360.1937
Concentration or Composition (by Analyte or Components) Aldosterone 1 mg/mL
Chemical Name or Material Aldosterone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.