missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aldosterone, 98%

Product Code. 10723411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723411 5 mg Glass bottle 5mg
10763724 25 mg Glass bottle 25mg
10195493 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10723411 Supplier Thermo Scientific Chemicals Supplier No. 215360050
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Aldosterone, CAS # 52-39-1, is a hormone secreted by the adrenal cortex, which regulates electrolyte and water balance by increasing the renal retention of sodium and potassium excretion.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Aldosterone is a pregnane-based steroidal hormone produced by the outer-section (zona glomerulosa) of the adrenal cortex in the adrenal gland by the action of aldosterone synthase (CYP11B2)
  • It modulates electrolyte and water balance by increasing the renal retention of sodium and potassium excretion.

Applications

  • Aldosterone binds to mineralocorticoid receptors (MR; NR3C2) and MR-aldosterone complexes regulating the expression of genes involved in the retention of sodium, secretion of potassium, and water reabsorption, resulting in an increase in the blood pressure
  • In vitro, it has been involved with the pathogenesis of obesity-related glomerulopathy through activation of Wnt/β-catenin signaling in podocytes
  • Aldosterone has been used in combination with salt to induce renal injury in mice
  • It is used to stimulate primary liver sinusoidal endothelial cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-39-1
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.45
MDL Number MFCD00051136
InChI Key PQSUYGKTWSAVDQ-ZVIOFETBSA-N
Synonym aldosterone, electrocortin, aldocortin, aldocorten, aldocortene, +-aldosterone, elektrocortin, d-aldosterone, reichstein x, 18-oxocorticosterone
PubChem CID 5839
ChEBI CHEBI:27584
IUPAC Name (8S,9S,10R,11S,13R,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-13-carbaldehyde
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4C(=O)CO)C=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Loss on Drying 6.0% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 mg
Beilstein 08, IV, 3491
Merck Index 15, 218
Specific Rotation +150° to +162° (20°C, 589nm) (c=0.1, CHCl3)
Formula Weight 360.45
Percent Purity ≥97.5% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aldosterone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.