missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Antibody (6F3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42925
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Aldolase C | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 6F3 | |
| Western Blot 1:5000 - 1:50000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| aldolase C, fructose-bisphosphate - (Uniprot# P09972) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 230 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction