missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Antibody (4A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25145SS
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal specifically detects Aldolase C in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Aldolase C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 | |
| P09972 | |
| ALDOC | |
| The N-terminal 20 amino acids of human Aldolase C (MPHSYPALSAEQKKELSDIA). [UniProt# P09972] | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human and Bovine. Other species have not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 4A9 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| Mouse | |
| 40 kDa | |
| 0.025 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 230 | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction