missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Antibody (4A9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25145B
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal specifically detects Aldolase C in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Aldolase C | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ALDOC | |
| The N-terminal 20 amino acids of human Aldolase C (MPHSYPALSAEQKKELSDIA). [UniProt# P09972] | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 230 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 4A9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction