missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Antibody (2E11), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59408G
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Aldolase C | |
| Monoclonal | |
| DyLight 488 | |
| ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
| Recombinant human Aldolase C (1-364aa) purified from E. coli | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 230 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 200000000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction