missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase B Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97910F
This item is not returnable.
View return policy
Description
Aldolase B Polyclonal specifically detects Aldolase B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Aldolase B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B | |
| Produced in rabbits immunized with E. coli-derived Human Aldolase B fragment. | |
| 0.1 mL | |
| Cancer | |
| 229 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction