missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aldolase A Recombinant Rabbit Monoclonal Antibody (24GB4710)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557329
This item is not returnable.
View return policy
Description
Aldolase A Recombinant Monoclonal Antibody for Western Blot
Aldolase encodes a member of the class I fructose-bisphosphate aldolase protein family. The encoded protein is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Mutations in this gene have been associated with Glycogen Storage Disease XII, an autosomal recessive disorder associated with hemolytic anemia. Disruption of this gene also plays a role in the progression of multiple types of cancers. Related pseudogenes have been identified on chromosomes 3 and 10.
Specifications
| Aldolase A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ALDOA | |
| ALDA; Aldo1; Aldo-1; ALDOA; Aldolase 1; aldolase 1, A isoform; aldolase A; aldolase A, fructose-bisphosphate; aldolase, fructose-bisphosphate A; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; fructose-bisphosphate aldolase A; GSD12; HEL-S-87p; Lung cancer antigen NY-LU-1; muscle-type aldolase; RNALDOG5; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11674, 226, 24189 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 24GB4710 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04075, P05064, P05065 | |
| ALDOA | |
| A synthesized peptide derived from human Aldolase (aa 7-20). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction