missing translation for 'onlineSavingsMsg'
Learn More

Aldo-keto Reductase 1C4/AKR1C4 Antibody [DyLight 650], Novus Biologicals™

Product Code. 30733535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733535 Supplier Novus Biologicals Supplier No. AF6957W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Aldo-keto Reductase 1C4/AKR1C4 Polyclonal antibody specifically detects Aldo-keto Reductase 1C4/AKR1C4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldo-keto Reductase 1C4/AKR1C4
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 3-alpha-HSD, AKR1C4, aldo-keto reductase family 1, member C4, C11, CDR, CHDR, DD4, DD-4, HAKRA
Host Species Sheep
Immunogen E. coli-derived recombinant human Aldo-keto Reductase 1C4/AKR1C4, Met1-Tyr323, Accession # NP_001809
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1109
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.