missing translation for 'onlineSavingsMsg'
Learn More

Aldo-keto Reductase 1C3/AKR1C3 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30734499 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734499 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734499 Supplier Novus Biologicals Supplier No. AF7678L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Aldo-keto Reductase 1C3/AKR1C3 Polyclonal antibody specifically detects Aldo-keto Reductase 1C3/AKR1C3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldo-keto Reductase 1C3/AKR1C3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 17-beta-hydroxysteroid dehydrogenase type 5, 3-alpha-HSD type 2, 3-alpha-hydroxysteroid dehydrogenase type 2, aldo-keto reductase family 1 member C3, aldo-keto reductase family 1, member C3 (3-alpha hydroxysteroid dehydrogenase, type II), Chlordecone reductase homolog HAKRb, DD-3, DD317-beta-HSD 5, DDH1, DDX, Dihydrodiol dehydrogenase 3, Dihydrodiol dehydrogenase type I, dihydrodiol dehydrogenase X, EC 1.1.1, EC 1.1.1.112,3-alpha-HSD type II, brain, EC 1.1.1.188, EC 1.1.1.213, EC 1.1.1.63, EC 1.1.1.64, EC 1.3.1.20, HA1753, HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase, HAKRe, HSD17B5, hydroxysteroid (17-beta) dehydrogenase 5, Indanol dehydrogenase, KIAA0119hluPGFS, PGFS, Prostaglandin F synthase, Testosterone 17-beta-dehydrogenase 5, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase
Host Species Sheep
Immunogen E. coli-derived recombinant human Aldo-keto Reductase 1C3/AKR1C3, Met1-Tyr323, Accession # P42330
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8644
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.