missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1C1/AKR1C1 Antibody (6D10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74042PCP
This item is not returnable.
View return policy
Description
Aldo-keto Reductase 1C1/AKR1C1 Monoclonal specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Aldo-keto Reductase 1C1/AKR1C1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| AKR1C1 | |
| Recombinant human AKR1C1 (1-323aa) purified from E. coli | |
| 100 μL | |
| metabolism | |
| 1645 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 6D10 | |
| Flow Cytometry | |
| 20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human AKR1C1 mAb, clone 6D10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human AKR1C1 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction