missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1B10/AKR1B10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | Aldo-keto Reductase 1B10/AKR1B10 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18224351
|
Novus Biologicals
NBP2-57972 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629026
|
Novus Biologicals
NBP2-57972-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldo-keto Reductase 1B10/AKR1B10 Polyclonal specifically detects Aldo-keto Reductase 1B10/AKR1B10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Aldo-keto Reductase 1B10/AKR1B10 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Lipid and Metabolism | |
| AKR1B11, AKR1B12, aldo-keto reductase family 1 member B10, aldo-keto reductase family 1, member B10 (aldose reductase), aldo-keto reductase family 1, member B11 (aldose reductase-like), Aldose reductase-like, aldose reductase-like 1, aldose reductase-like peptide, Aldose reductase-related protein, ALDRLn, ARL1, ARL-1SI reductase, ARP, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.21, hARP, HIS, HSI, MGC14103, Small intestine reductase | |
| AKR1B10 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 57016 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FKLSDEEMATILSFNRNWRACNVLQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title