missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1B10/AKR1B10 Antibody (1A6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057016-M01
This item is not returnable.
View return policy
Description
Aldo-keto Reductase 1B10/AKR1B10 Monoclonal antibody specifically detects Aldo-keto Reductase 1B10/AKR1B10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| Aldo-keto Reductase 1B10/AKR1B10 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AKR1B11, AKR1B12, aldo-keto reductase family 1 member B10, aldo-keto reductase family 1, member B10 (aldose reductase), aldo-keto reductase family 1, member B11 (aldose reductase-like), Aldose reductase-like, aldose reductase-like 1, aldose reductase-like peptide, Aldose reductase-related protein, ALDRLn, ARL1, ARL-1SI reductase, ARP, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.21, hARP, HIS, HSI, MGC14103, Small intestine reductase | |
| AKR1B10 (NP_064695, 76 a.a. ∽ 143 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VSKLWPTFFERPLVRKAFEKTLKDLKLSYLDVYLIHWPQGFKSGDDLFPKDDKGNAIGGKATFLDAWE | |
| 0.1 mg | |
| Cancer, Lipid and Metabolism | |
| 57016 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 1A6 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| NP_064695 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction