Learn More
ALDH9A1 Mouse anti-Human, CoraLite 594, Clone: 3C5C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67372-100UL
Description
The ALDH9A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALDH9A1. This antibody recognizes human antigen. The ALDH9A1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| ALDH9A1 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 223 | |
| IgG1 |
| Immunofluorescence | |
| 3C5C9 | |
| Mouse | |
| Primary | |
| Human |