missing translation for 'onlineSavingsMsg'
Learn More

ALDH9A1 Antibody (3C6), Novus Biologicals™

Product Code. 18341358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341358 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341358 Supplier Novus Biologicals Supplier No. H00000223M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALDH9A1 Monoclonal antibody specifically detects ALDH9A1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH9A1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3C6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000687
Gene Alias aldehyde dehydrogenase 9 family, member A1, Aldehyde dehydrogenase E3 isozyme, Aldehyde dehydrogenase family 9 member A1, ALDH4, ALDH7, ALDH9aldehyde dehydrogenase (NAD+), E34-trimethylaminobutyraldehyde dehydrogenase, EC 1.2.1, EC 1.2.1.19, EC 1.2.1.3, EC 1.2.1.47, EC 1.2.1.8, Gamma-aminobutyraldehyde dehydrogenase, R-aminobutyraldehyde dehydrogenase, TMABADH
Host Species Mouse
Immunogen ALDH9A1 (NP_000687, 173 a.a. ~ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CGNAMVFKPSPFTPVSALLLAEIYSEAGVPPGLFNVVQGGAATGQFLCQHPDVAKVSFTGSVPTGMKIMEMSAKGIKPVTLELGGKSPLIIFSDCDMN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 223
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.