missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ALDH7A1 Polyclonal antibody specifically detects ALDH7A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ALDH7A1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: EVFAWNNEVKQGLSSSIFTKDLGRIFRWLGPKGSDCGIVNVNIPTSGAEIgGAFGGEKHTGGGRESGSDAWKQYMRRSTCTINY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?