missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH7A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554750
This item is not returnable.
View return policy
Description
Immunogen sequence: DLSLVVPSAL FAAVGTAGQR CTTARRLFIH ESIHDEVVNR LKKAYAQIRV GNPWDPNVLY GPLHTKQAVS MFLGAVEEAK KEGGTVVYGG KVMDRPGNYV EPTIVTGLGH DASIAHTETF APILY Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.
Specifications
| ALDH7A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH7A1 | |
| 26g turgor protein homolog; Ald7a1; aldehyde dehydrogenase 7 family member A1; aldehyde dehydrogenase 7 family, member A1; aldehyde dehydrogenase family 7 member A1; aldehyde dehydrogenase family 7, member A1; Aldh7a1; Alpha-AASA dehydrogenase; alpha-aminoadipic semialdehyde dehydrogenase; antiquitin; Antiquitin 1; Antiquitin1; Antiquitin-1; ATQ1; Betaine aldehyde dehydrogenase; D18Wsu181e; delta1-piperideine-6-carboxylate dehydrogenase; delta1-piperideine-6-carboxylate dehydrogenease; EPD; P6c dehydrogenase; PDE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 501 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49419 | |
| ALDH7A1 | |
| Recombinant protein corresponding to Human ALDH7A1. Recombinant protein control fragment (Product #RP-93154). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction