missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH7A1 Antibody (SR1977), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £358.00
Specifications
| Antigen | ALDH7A1 |
|---|---|
| Clone | SR1977 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620734
|
Novus Biologicals
NBP3-21975-100ul |
100 μg |
£358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619903
|
Novus Biologicals
NBP3-21975-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH7A1 Monoclonal antibody specifically detects ALDH7A1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| ALDH7A1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1 | |
| A synthesized peptide derived from human ALDH7A1 (Uniprot #: P49419) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1977 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 501 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title