missing translation for 'onlineSavingsMsg'
Learn More

ALDH7A1 Antibody (101), DyLight 755, Novus Biologicals™

Product Code. 18326951 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326951 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326951 Supplier Novus Biologicals Supplier No. NBP289926IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ALDH7A1 Monoclonal specifically detects ALDH7A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH7A1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 101
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ALDH7A1 (P49419-2; Ser2-Gln511).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 501
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.