missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH6A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20452-1-AP-20UL
This item is not returnable.
View return policy
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids.Specifications
| ALDH6A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH6A1 | |
| ALDH6A1, MMSADHA, MMSDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104776, 4329, 81708 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02252, Q02253, Q9EQ20 | |
| ALDH6A1 | |
| ALDH6A1 Fusion Protein Ag13879 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction