missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH6A1 Monoclonal Antibody (147CT8.3.4)
Description
ALDH6A1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids.
Specifications
Specifications
| Antigen | ALDH6A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 147CT8.3.4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ALDH6A1 |
| Gene Accession No. | Q02252 |
| Gene Alias | 1110038I05Rik; AI314632; aldehyde dehydrogenase 6 family member A1; aldehyde dehydrogenase 6 family, member A1; aldehyde dehydrogenase family 6 member A1; aldehyde dehydrogenase family 6, subfamily A1; Aldh6a1; malonate-semialdehyde dehydrogenase; methylmalonate semialdehyde dehydrogenase; methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; MGC40271; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSADHA; Mmsdh; testicular tissue protein Li 122 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?