missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH5A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583101
This item is not returnable.
View return policy
Description
Immunogen sequence: RKWYNLMIQN KDDLARIITA ESGKPLKEAH GEILYSAFFL EWFSEEARRV YGDIIHTPAK DRRALVLKQP IGVAAVITPW NFPSAM Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 87%.
Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
Specifications
| ALDH5A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH5A1 | |
| 6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51649 | |
| ALDH5A1 | |
| Recombinant protein corresponding to Human ALDH5A1. Recombinant protein control fragment (Product #RP-94682). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction