missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH5A1 Antibody (17F5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59409AF647
This item is not returnable.
View return policy
Description
ALDH5A1 Monoclonal antibody specifically detects ALDH5A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| ALDH5A1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| aldehyde dehydrogenase 5 family, member A1, EC 1.2.1, EC 1.2.1.24, NAD(+)-dependent succinic semialdehyde dehydrogenase, SSADHmitochondrial | |
| Recombinant human ALDH5A1 (48-535aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Neuroscience | |
| 7915 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| 17F5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction