Learn More
Invitrogen™ ALDH4A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596180
Description
Immunogen sequence: LYVPHSLWPQ IKGRLLEEHS RIKVGDPAED FGTFFSAVID AKSFARIKKW LEHARSSPSL TILAGGKCDD SVGYFVEPCI VESKDPQEPI MKEEIFGPVL SVYVYPDDKY KETLQLVDST TSYGLTGAVF SQDKDVVQEA TKVLRNAAGN FYINDKSTGS IVGQQPFGGA RASGTNDKPG GPHYILRWTS PQVIKETHKP LGDWSYAYMQ; Positive Samples: HepG2, K-562, A-549, BT-474, Mouse skeletal muscle, Mouse intestine, Mouse kidney; Cellular Location: Mitochondrion matrix.
ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
Specifications
| ALDH4A1 | |
| Polyclonal | |
| Unconjugated | |
| Aldh4a1 | |
| A930035F14Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 4 family member A1; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; aldehyde dehydrogenase family 4 member A1; ALDH4; aldh4a1; Aldh5a1; delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; E330022C09; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5cd; P5CDh; P5cdhl; P5cdhs; RP11-128M10.1; similar to aldehyde dehydrogenase 4 family, member A1; Ssdh1; zgc:63592 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 212647, 8659 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 6.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30038, Q8CHT0 | |
| Aldh4a1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 354-563 of human ALDH4A1 (NP_0037392). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.