missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH4A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54739
This item is not returnable.
View return policy
Description
ALDH4A1 Polyclonal specifically detects ALDH4A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ALDH4A1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| aldehyde dehydrogenase 4 family, member A1, Aldehyde dehydrogenase family 4 member A1, ALDH4DKFZp779M035, delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial, EC 1.5.1.12, mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase, P5C dehydrogenase, P5CD, P5CDh | |
| Rabbit | |
| 24 kDa | |
| 100 μL | |
| Primary | |
| Bovine: 86%; . | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q5TF55 | |
| ALDH4A1 | |
| Synthetic peptides corresponding to ALDH4A1(aldehyde dehydrogenase 4 family, member A1) The peptide sequence was selected from the N terminal of ALDH4A1. Peptide sequence QGKTVIQAEIDAAAELIDFFRFNAKYAVELEGQQPISVPPSTNSTVYRGL. | |
| Protein A purified | |
| RUO | |
| 8659 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction