missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH3B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15746-1-AP-20UL
This item is not returnable.
View return policy
Description
ALDH3B2 (aldehyde dehydrogenase 3 family, member B2), also known as ALDH8, is a 385 amino acid protein that belongs to the ALDH family and is involved in the pathway of alcohol metabolism. Expressed in salivary gland tissue, ALDH3B2 functions to catalyze the NADP+-dependent conversion of an aldehyde into an acid. The gene encoding ALDH3B2 maps to human chromosome 11, which houses over 1, 400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.Specifications
| ALDH3B2 | |
| Polyclonal | |
| Unconjugated | |
| ALDH3B2 | |
| Aldehyde dehydrogenase 8, ALDH3B2, ALDH8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 222, 621603, 688800 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9Q3E1, P48448 | |
| Aldh3b2 | |
| ALDH3B2 Fusion Protein Ag8197 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction