missing translation for 'onlineSavingsMsg'
Learn More

ALDH3B2 Antibody (3E6), Novus Biologicals™

Product Code. 18360168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360168 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360168 Supplier Novus Biologicals Supplier No. H00000222M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALDH3B2 Monoclonal antibody specifically detects ALDH3B2 in Human samples. It is validated for ELISA, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3B2
Applications ELISA, Protein Array
Classification Monoclonal
Clone 3E6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH07685
Gene Alias acetaldehyde dehydrogenase 8, aldehyde dehydrogenase 3 family, member B2, Aldehyde dehydrogenase 8, ALDH8aldehyde dehydrogenase family 3 member B2, EC 1.2.1, EC 1.2.1.5
Host Species Mouse
Immunogen ALDH3B2 (AAH07685, 1 a.a. ~ 385 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKDEPRSTNLFMKLDSVFIWKEPFGLVLIIAPWNYPLNLTLVLLVGALAAGSCVVLKPSEISQGTEKVLAEVLPQYLDQSCFAVVLGGPQETGQLLEHKLDYIFFTGSPRVGKIVMTAATKHLTPVTLELGGKNPCYVDDNCDPQTVANRVAWFCYFNAGQTCVAPDYVLCSPEMQERLLPALQSTITRFYGDDPQSSPNLGRIINQKQFQRLRALLGCGRVAIGGQSNESDRYIAPTVLVDVQETEPVMQEEIFGPILPIVNVQSVDEAIKFINWQEKPLALYAFSNSSQVVNQMLERTSSGSFGGNEGFTYISLLSVPFGGVGHSGMGRYHGKFTFDTFSHHRTCLLAPSGLEKLKEIHYPPYTDWNQQLLRWGMGSQSCTLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 222
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.