missing translation for 'onlineSavingsMsg'
Learn More

ALDH3B1 Antibody (1F2), Novus Biologicals™

Product Code. 18394428 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394428 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394428 Supplier Novus Biologicals Supplier No. H00000221M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALDH3B1 Monoclonal antibody specifically detects ALDH3B1 in Human samples. It is validated for ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH3B1
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1F2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13584
Gene Alias aldehyde dehydrogenase 3 family, member B1, aldehyde dehydrogenase 3B1, Aldehyde dehydrogenase 7, aldehyde dehydrogenase family 3 member B1, ALDH4, ALDH7FLJ34710, EC 1.2.1, EC 1.2.1.5, FLJ26433
Host Species Mouse
Immunogen ALDH3B1 (AAH13584, 1 a.a. ~ 468 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDPLGDTLRRLREAFHAGRTRPAEFRAAQLQGLGRFLQENKQLLHDALAQDLHKSAFESEVSEVAISQGEVTLALRNLRAWMKDERVPKNLATQLDSAFIRKEPFGLVLIIAPWNYPLNLTLVPLVGALAAGNCVVLKPSEISKNVEKILAEVLPQYVDQSCFAVVLGGPQETGQLLEHRFDYIFFTGSPRVGKIVMTAAAKHLTPVTLELGGKNPCYVDDNCDPQTVANRVAWFRYFNAGQTCVAPDYVLCSPEMQERLLPALQSTITRFYGDDPQSSPNLGRIINQKQFQRLRALLGCGRVAIGGQSDESDRYIAPTVLVDVQEMEPVMQEEIFGPILPIVNVQSLDEAIEFINRREKPLALYAFSNSSQVVKRVLTQTSSGGFCGNDGFMHMTLASLPFGGVGASGMGRYHGKFSFDTFSHHRACLLRSPGMEKLNALRYPPQSPRRLRMLLVAMEAQGCSCTLL
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.