missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ALDH3A2 Recombinant Protein

Brand:  Abnova™ H00000224-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191471

  • £425.00 / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH02430
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
81.62
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ALDH3A2
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
224
ALDH3A2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ALDH10/DKFZp686E23276/FALDH/FLJ20851/SLS
ALDH3A2
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers