missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH1L2 Polyclonal Antibody

Description
Immunogen sequence: KCGGLQLQNE DVYMATKFEG FIQKVVRKLR GEDQEVELVV DYISKEVNEI MVKMPYQCFI NGQFTDADDG KT Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 79%.
This gene encodes a member of both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. This member is the mitochondrial form of 10-formyltetrahydrofolate dehydrogenase (FDH), which converts 10-formyltetrahydrofolate to tetrahydrofolate and CO2 in an NADP(+)-dependent reaction, and plays an essential role in the distribution of one-carbon groups between the cytosolic and mitochondrial compartments of the cell. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
| Antigen | ALDH1L2 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ALDH1L2 |
| Gene Accession No. | Q3SY69 |
| Gene Alias | 10-formyltetrahydrofolate dehydrogenase ALDH1L2; aldehyde dehydrogenase 1 family member L2; aldehyde dehydrogenase 1 family, member L2; aldehyde dehydrogenase 1L2; Aldehyde dehydrogenase family 1 member L2; aldehyde dehydrogenase family 1 member L2, mitochondrial; ALDH1L2; D330038I09Rik; mitochondrial 10-formyltetrahydrofolate dehydrogenase; Mitochondrial 10-FTHFDH; mtFDH; probable 10-formyltetrahydrofolate dehydrogenase ALDH1L2 |
| Gene Symbols | ALDH1L2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?