missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH1L1 Monoclonal Antibody (2E7)
Mouse Monoclonal Antibody
£430.00 - £1482.00
Specifications
| Antigen | ALDH1L1 |
|---|---|
| Clone | 2E7 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17805107
|
Invitrogen™
MA547305 |
500 μL |
£1482.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17815167
|
Invitrogen™
MA547403 |
100 μL |
£430.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH1L1 Monoclonal Antibody for Western Blot, ICC/IF
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
| ALDH1L1 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| O75891, P28037, Q8R0Y6 | |
| 107747, 10840, 64392 | |
| Amino acids 402-902 of human ALDH1L1 protein expressed in and purified from E. coli. | |
| Antibody |
| 2E7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat | |
| ALDH1L1 | |
| 10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2 | |
| ALDH1L1 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title