missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22220-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to ALDH1B1.
Aldehyde dehydrogenases (ALDHs) mediate NADP+-dependent oxidation of aldehydes into acids during detoxification of alcohol-derived acetaldehyde, lipid peroxidation and metabolism of corticosteroids, biogenic amines and neurotransmitters. Alcohol drinking habits and cardiovascular disease risk factors may be associated with ALDH gene variants. ALDH1B1 (Aldehyde dehydrogenase family 1 member B1), also known as ALDH5 or ALDHX (Aldehyde dehydrogenase X, mitochondrial), is a 517 amino acid mitochondrial protein that is expressed in the liver, testis and to a lesser extent in brain. ALDH1B1 belongs to the aldehyde dehydrogenase family and may play a major role in ethanol detoxification.Specifications
| ALDH1B1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH1B1 | |
| Aldehyde dehydrogenase 5, ALDH1B1, ALDH5, ALDHX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219, 298079, 72535 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30837, Q66HF8, Q9CZS1 | |
| Aldh1b1 | |
| ALDH1B1 Fusion Protein Ag17557 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction