missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£225.00
Specifications
| Antigen | ALDH1A3 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:3000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000. Use reported by customer review, Immunohistochemistry-Paraffin 1:100 - 1:1000, Immunohistochemistry-Frozen reported by customer review and in scientific literature (PMID 26713822), Immunoblotting reported in scientific literature (PMID 27572106) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
ALDH1A3 Polyclonal specifically detects ALDH1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
| ALDH1A3 | |
| Western Blot 1:500 - 1:3000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000. Use reported by customer review, Immunohistochemistry-Paraffin 1:100 - 1:1000, Immunohistochemistry-Frozen reported by customer review and in scientific literature (PMID 26713822), Immunoblotting reported in scientific literature (PMID 27572106) | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS with 0.02% Sodium Azide | |
| aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 | |
| ALDH1A3 | |
| IgG | |
| Affinity Purified | |
| 56 kDa |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P47895 | |
| 220 | |
| Recombinant protein encompassing a sequence within the center region of human ALDH1A3. The exact sequence is proprietary. | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title